Anti-Bckdk Rabbit Polyclonal Antibody
Catalog # 76067-256
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:[3-methyl-2-oxobutanoate dehydrogenase [lipoamide]] kinase, mitochondrial
- Antigen Symbol:Bckdk
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:400 μL
- Epitope:159-188 aa
- Form:Liquid
- Gene ID:12041
- Modification:None
- Molecular Weight:47 kDa
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0.48 mg/ml
- Shipping Temperature:4°C
- Immunogen:This Mouse Bckdk antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 159-188 amino acids from the Central region of mouse Bckdk.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Cat. No.:76067-256
Specifications
About this item
For WB starting dilution is: 1:1000
Type: Primary
Antigen: Bckdk
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 159-188
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse