ImmunO™ Sheep Serum, MP Biomedicals
Catalog # IC642951
Supplier: MP Biomedicals
Specifications
- Serum format:Lyophilized
- Description:Normal Sheep Serum
- Size:50 mL
- Storage Temperature:2 - 8 °C
- Cat. No.:IC642951
- Supplier no.:642951
Specifications
About this item
Sheep serum is useful for the propagation of long-term cultures of erythrocytic stages of the Theileria uilenbergi intracellular protozoan parasite.
- Sterile filtered
Reconstitute with 50 mL of sterile distilled water.