Order Entry
Puerto Rico
ContactUsLinkComponent
ImmunO™ Mouse Serum, MP Biomedicals
ImmunO™ Mouse Serum, MP Biomedicals
Catalog # IC642931
Supplier:  MP Biomedicals
undefined
ImmunO™ Mouse Serum, MP Biomedicals
Catalog # IC642931
Supplier:  MP Biomedicals
Supplier Number:  08642931

Specifications

  • Serum format:
    Lyophilized
  • Description:
    Normal Mouse Serum
  • Size:
    10 mL
  • Storage Temperature:
    2 - 8 °C
  • Cat. No.:
    IC642931
  • Supplier no.:
    642931

Specifications

About this item

Mouse serum is used for blocking agents and controls in immunoassays

  • Sterile filtered

Reconstitute with 10.0 mL of distilled water.

Mouse Serum is a pooled preparation obtained from a normal mouse population.