Order Entry
Puerto Rico
ContactUsLinkComponent
ImmunO™ Goat Serum, MP Biomedicals
ImmunO™ Goat Serum, MP Biomedicals
Catalog # IC642921
Supplier:  MP Biomedicals
undefined
ImmunO™ Goat Serum, MP Biomedicals
Catalog # IC642921
Supplier:  MP Biomedicals
Supplier Number:  08642921

Specifications

  • Serum format:
    Lyophilized
  • Description:
    Normal Goat Serum
  • Size:
    50 mL
  • Storage Temperature:
    2 - 8 °C
  • Cat. No.:
    IC642921
  • Supplier no.:
    642921

Specifications

About this item

Goat serum can be used in immunocytochemistry and blocking.

  • Sterile filtered