$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Buffers
Assay Buffers
Buffer P3 (Yellow), Zymo Research
Buffer P3 (Yellow), Zymo Research
:
Zymo Research
Buffer P3 (Yellow), Zymo Research
:
Zymo Research
Brand Name:
Zymo Research