Vial Racks for Cetec Autosampler Vials, PerkinElmer
Supplier: PerkinElmer
Certificates
About this item
These polypropylene racks accommodate 40 headspace auto-sampler vials.
- Autoclavable and stackable
- Good chemical and temperature resistance
- Features end plates for labeling and numbering for tube identification