

Acetic acid ≥99.8% (by GC) eluent additive, for LC-MS, Fluka™
Catalog # BJ49199-50ML
Supplier: Honeywell Research Chemicals
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle
- Size:50 ml
- Cat. No.:BJ49199-50ML
- Supplier no.:49199-50ML
Specifications
Specification Test Results
- Vapor Density:2.07 (vs air)
- Assay:99.5 - 100.5 %
- Purity (GC):Min. 99.8 %
- Solidification Point:16.3 - 16.7 °C
- Water (Karl Fischer):Max. 0.2 %
- Silver (Ag):Max. 0.01 ppm
- Aluminium (Al):Max. 0.05 ppm
- Arsenic (As):Max. 0.01 ppm
- Barium (Ba):Max. 0.01 ppm
- Beryllium (Be):Max. 0.01 ppm
- Bismuth (Bi):Max. 0.1 ppm
- Calcium (Ca):Max. 0.2 ppm
- Cadmium (Cd):Max. 0.02 ppm
- Cobalt (Co):Max. 0.01 ppm
- Chromium (Cr):Max. 0.05 ppm
- Copper (Cu):Max. 0.01 ppm
- Iron (Fe):Max. 0.2 ppm
- Germanium (Ge):Max. 0.05 ppm
- Potassium (K):Max. 0.1 ppm
- Lithium (Li):Max. 0.01 ppm
- Magnesium (Mg):Max. 0.1 ppm
- Manganese (Mn):Max. 0.01 ppm
- Molybdenum (Mo):Max. 0.02 ppm
- Sodium (Na):Max. 0.5 ppm
- Nickel (Ni):Max. 0.05 ppm
- Lead (Pb):Max. 0.02 ppm
- Strontium (Sr):Max. 0.01 ppm
- Titanium (Ti):Max. 0.01 ppm
- Thallium (Tl):Max. 0.05 ppm
- Vanadium (V):Max. 0.01 ppm
- Zinc (Zn):Max. 0.05 ppm
- Zirconium (Zr):Max. 0.1 ppm
- Chloride (Cl):Max. 0.5 ppm
- Phosphate (PO₄):Max. 0.5 ppm
- Sulfate (SO₄):Max. 0.5 ppm
- Suitability for the LC-MS:complying
- Acetic aNHydride (GC):Max. 0.01 %
- APHA:Max. 10
- Explosion Limit:16 %, 92 °F
- Refractive Index:n20/D 1.371 (lit.)
Specification Test Results
About this item
:
:60.05 g/mol
Boiling Pt:118 °C (1.013 hPa)
Melting Pt:16 °C
Density:1.05 g/cm³ (20 °C)
Flash Pt:40 °C
Storage Temperature:Ambient
EINECS:200-580-7
UN:2789
ADR:8,II
:GHS02
:GHS05
CAS Number:64-19-7
Formula:H₃CCOOH
MDL Number:MFCD00036152