KF Streptococcal Agar Base Dehydrated Culture Media, HiMedia Laboratories
95021-562
: HiMedia
:
- Description:KF Streptococcal Agar Base
- Cat. No.:95021-562
- Supplier no.:M248-500G
For the selective isolation and enumeration of fecal streptococci in surface water by direct plating or by membrane filter method.
- Microbiological culture media
- 500 g poly bottles