Red Blood Cell Lysis Buffer, 1X, BioVision
76020-790
: Avantor
:
- Description:Cell Lysis Buffer
- Size:100 ml
- Brand Name:BioVision Inc.
- Cat. No.:76020-790
Buffer for efficient removal of RBCs from whole blood, tissues or tumor cells.
- Ready to use as supplied.
- Simple & convenient method for highly efficient removal of Red Blood cells from whole blood, tissues or tumor cells.
- The reagent does not affect leukocytes, normal tissue, or tumor cells and does not interfere with down-stream functional assays.
Red Blood Cell Lysis buffer for quick removal of RBC from whole blood, tissues and tumor cells.