Low Temperature PCR Racks
Supplier: Heathrow Scientific
Certificates
About this item
These Low Temperature PCR Racks feature a sliding lid that roates 180° to become a convenient stand that the rack sits on. These PCR racks also feature ergonomic textured finger grips that provide a secure hold, reducing the chance of dropping the rack, and makes handling and transportation easy.
- Polycarbonate lid serves as cover and elevated rack base
- Lid can be used in right or left-handed orientation and is detachable
- Sliding lid rotates 180° to become a convenient stand that the rack sits on
- Side opening enables faster cooling of samples to ensure integrity
- Stackable design saves valuable bench-top and freezer space
- Features alphanumeric identification for quick location and retrieval of tubes
- Can be used for freezer storage down to −80 °C
The lids have a side opening that enables faster cooling of samples to ensure integrity and are made from translucent polycarbonate for easy sample and well identification. The base of the rack is made from robust ABS construction that helps keep samples safe, so you don't have to spend extra time and money on expensive retesting. This rack comes in multiple colors for coding options in the lab.