Azospirillum Medium with 0.17% Agar, Twin Pack, Dehydrated Culture Media, HiMedia
Catalog # 95021-934
Supplier: HiMedia
CAS Number:
Specifications
- Description:Azospirillum Medium with 0.17% Agar, Twin Pack
- Cat. No.:95021-934
- Supplier no.:M518-500G
Specifications
About this item
For the cultivation of Azospirillum species
- Microbiological culture media
- Packaged in 500 g poly bottles