ContactUsLinkComponent
Cellular Senescence Activity Assay, Enzo Life Sciences
Cellular Senescence Activity Assay, Enzo Life Sciences
  76003-296
 :  
undefined
Cellular Senescence Activity Assay, Enzo Life Sciences
  76003-296
 :  KIT129-0120
 :  
  • Cat. No.:
    76003-296
  • No. of assays/kit:
    120 assays
Highly reproducible SA-β-gal activity kit
  • Quantify SA-β-gal activity in a fluorescence plate reader
  • Easy-to-use protocol with results in less than 90 minutes
  • Fully compatible with all cell types
For the determination of cellular senescence by measuring SA-β-gal activity using a fluorometric substrate. The 96-well Cellular senescence assay kit provides an easy-to-use and efficient method to determine the cellular senescence by measuring SA-β-gal activity using a fluorometric substrate. This quantitative assay uses cell lysate for both SA-β-galactosidase activity determination and normalization of samples containing different cell numbers. Each kit provides sufficient quantities to perform up to 120 assays in a 96-well plate.
 : Contains 2X Cell lysis buffer, 2X Reaction buffer, SA-?-gal substrate (20X), Stop solution.