Oxytocin ELISA kit, Enzo Life Sciences
Catalog # 76002-268
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Competitive
- Size:Kit
- Sensitivity:15 pg/mL (Range 15.6 - 1000 pg/mL)
- Cat. No.:76002-268
- No. of tests:5× 96 wells
Specifications
About this item
The most sensitive, specific and flexible alternative to more costly and time-consuming methods for oxytocin detection.
- Highly specific ELISA which does not detect vasopressin, providing confidence in assay results
- Ultra-sensitive measurement, detecting as little as 15 pg/mL oxytocin
- Cost effective solution, faster and less costly than LC/MS
- Easy-to-use liquid color-coded reagents and pre-coated plates minimizes errors
- Widely published in peer reviewed literature
- Save with bulk kit package!
For the quantitative determination of Oxytocin in culture supernatants, milk, plasma, and serum from any species. Validated sample types include cerebral spinal fluid, saliva, tissue and urine. The Oxytocin EIA kit is a colorimetric competitive enzyme immunoassay kit with results overnight + 1 hour. Absorbance is read at 405 nm. Very low reactivity with vasopressin provides confidence in assay results. Sensitive quantification of 15 pg/mL oxytocin. Ready-to-use liquid color-coded reagents reduce errors in the lab.
Ordering information: Contains GxR Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: ADI-900-153A (1 x 96 wells).