Arg8-Vasopressin ELISA Kit, Enzo Life Sciences
Catalog # 76002-264
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Competitive
- Sensitivity:2.84 pg/mL (Range 4.10 - 1000 pg/mL)
- Cat. No.:76002-264
- No. of tests:96 wells
Specifications
About this item
Highly sensitive ELISA kit for the detection of Arg8-Vasopressin in serum, plasma and tissue culture media.
- High sensitivity, detecting as little as 2.84 pg/mL
- Highly specific with negligible reactivity with oxytocin
- Simple and easy-to-use liquid color-coded reagents reduces error
- Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of Arg8-Vasopressin in human serum, human plasma, and tissue culture media.
The Arg8-Vasopressin EIA kit is a colorimetric competitive enzyme immunoassay kit with results overnight + 1 hour. Absorbance is read at 450 nm.
Ordering information: Contains GxR IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, SA-HRP, TMB Substrate, Stop solution. Alternative size available: ADI-901-017A (5 x 96 wells).