Specifications
- Description:TRUBLU Urine and Serum Pregnancy Test Kit
- Cat. No.:103388-942
Specifications
About this item
TRUBLU Urine and Serum hCG Pregnancy Test Kits are capable of detecting pregnancy as early as the first day of the first missed menses.
- Sensitivity of 25 mlU/m
- Results in three minutes for urine and five minutes for serum
- CLIA waived for urine testing
- Made in the USA
The TRUBLU Urine and Serum hCG Pregnancy test kit has a 25 mIU/m sensitivity, and is capable of detecting pregnancy as a early as one day after missed menses. With rapid results, in three minutes for urine and five minutes for serum, this kit offers quality and dependability.