Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Lead
Lead (Test), granulated silver-free
Lead (Test), granulated silver-free
76338-316
:
ATLANTIC SCHMELZTIEGEL
:
Lead (Test), granulated silver-free
76338-316
:
ATLANTIC SCHMELZTIEGEL
:
10607-20KG
:
Size:
20 kg
Environmentally Preferable:
Cat. No.:
76338-316
Silver:
≤0.4 ppm
:
:
207.2 g/mol
:
7439-92-1
:
Pb
:
MFCD00134050
:
1740
:
327
:
12,05410