Order Entry
United States
ContactUsLinkComponent
CYGREEN® Nucleic Acid Dye, Enzo
CYGREEN® Nucleic Acid Dye, Enzo
Catalog # 76003-286
Supplier:  Enzo Life Sciences
CAS Number:  
CYGREEN® Nucleic Acid Dye, Enzo
Catalog # 76003-286
Supplier:  Enzo Life Sciences
Supplier Number:  GEN105-0100
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    CYGREEN® Nucleic Acid Dye
  • Size:
    100 µl
  • Environmentally Preferable:
  • Cat. No.:
    76003-286

Specifications

About this item

Nucleic acid dye for gel staining and higher sensitivity qPCR.

  • Higher sensitivity allows detection of low target concentration
  • Greater PCR efficiency for accurate and reproducible results
  • Soluble in water for higher stability
  • Compatible with various enzyme systems
  • Suitable for commonly used rotor-type and plate-type qPCR platforms
  • Validated for qPCR applications and gel staining
  • Lower cost per assay compared to commonly used dyes

CYGREEN® Nucleic acid dye is a DNA intercalating agent that is used to stain DNA. The DNA-dye complex emits a fluorescence spectra that makes it suitable for qPCR and gel staining applications.CYGREEN® Nucleic acid dye has excitation/emission spectra similar to commonly used qPCR dyes and can be substituted in to increase sensitivity and reduce cost.