BIO-PROBE® Lambda Probe, Enzo Life Sciences
BIO-PROBE® Lambda Probe, 2 µg
Catalog # 76002-526
Supplier: Enzo Life Sciences
Supplier #: ENZ-40840
BIO-PROBE® Lambda Probe, Enzo Life Sciences
Catalog # 76002-526
Supplier: Enzo Life Sciences
CAS Number:
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Negative control probe for in situ hybridization.
The Lambda BIO-PROBE® labeled probe is prepared by nick translation of the lambda bacteriophage strain c1857ind1 Sam7. Fragment size range: 100-1000 base pairs (as estimated by agarose gel electrophoresis).
Specifications
Specifications
- Description:BIO-PROBE® Lambda Probe
- Size:2 µg
- Cat. No.:76002-526
- Formulation:Liquid. In 10 mM TRIS-HCl, pH 7.5, containing 1mM EDTA
- Concentration:20 µg/mL
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



