Order Entry
Puerto Rico
ContactUsLinkComponent
Accredo Safety Lockout Tagout Hasp 12 Hole, ZING Enterprises
Accredo Safety Lockout Tagout Hasp 12 Hole, ZING Enterprises
Catalog # 76026-318
Supplier:  ZING Enterprises
undefined
Accredo Safety Lockout Tagout Hasp 12 Hole, ZING Enterprises
Catalog # 76026-318
Supplier:  ZING Enterprises
Supplier Number:  7240

Specifications

  • Description:
    Standard Steel Hasp -12 Hole
  • Length:
    8.5"
  • Material:
    Steel
  • Cat. No.:
    76026-318
  • Width:
    2.25"

Specifications

About this item

Lockout tagout hasp.

  • Supports OSHA Standard 1910.147
  • High quality
  • Very durable

12 Hole Lockout Hasp is designed to secure switches, valves and other energy devices from use while equipment is being serviced or repaired. The hasp is made from durable coated steel and measures 8.5’’L x 2.25’’W. The closed hasp hole size measures 1 ½’’. The hasp will accommodate 12 padlocks with a max shackle diameter of 3/8’’.

Certifications: OSHA 1910.147