Wright-Giemsa stain
Product Details & Documents
Used in Prussian Blue Stain for Non-Hemoglobin Iron Sundberg and Broman (1955). Stain results - Particulate Iron and Non-granular cytoplasm of many Macrophages: Blue to Blue-green Other: Comparable to original films stained.
Clark, G.: Staining Procedures, Williams & Wilkins Co. Baltimore, 3rd Ed., c.1973, p.130. Sunberg,. R.D. and Broman, H., Blood, 10160-6, 1955.
- MDL Number:MFCD00132593
Specifications
Specifications
- Size:500 ml