ContactUsLinkComponent
Sterile Spencer Stitch Scissors, Disposable, Sklar
Sterile Spencer Stitch Scissors, Disposable, Sklar
Catalog # 75996-386
Supplier:  Sklar
undefined
Sterile Spencer Stitch Scissors, Disposable, Sklar
Catalog # 75996-386
Supplier:  Sklar
Supplier Number:  96-2596M
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Length:
    114 mm (4½")
  • Curvature:
    Straight
  • Cat. No.:
    75996-386
Specifications
About this item
Merit™ Sterile Spencer Stitch Scissors are primarily used for suture removal.
  • Small hook-shaped tip on one blade
  • Delicate tips for small suture removal
  • Ready-to-use
These scissors have a small hook shaped tip on one blade that slides under sutures to slightly lift them before cutting for removal. This hook also holds the suture so it does not slip off the blade of the scissors prior to cutting. They are most commonly available in lengths of 3-1/2" and 4-1/2" and therefore typically have smaller, more delicate tips that are used for removing smaller sutures. They should not be used for cutting tissue. Merit™ Sterile instruments are ready-to-use, sterile, disposable, office-grade instruments sold in bulk quantities.