MicroSeal® VP-Sealed Edge Flat Mop Wiper, Berkshire
Catalog # 76027-820
Supplier: Berkshire
Specifications
- Description:MicroSeal® VP-Sealed Edge Flat Mop Wiper
- Material:Knitted Polyester
- Packed:Bulk
- Cat. No.:76027-820
- Details:Ultrasonically Sealed
Specifications
About this item
The MicroSeal® VP- Mop Wiper is a sealed edge, laundered, knit wiper.
- Lightweight
- Tight knit construction
- Low abrasion
It is composed of 100% continuous filament polyester knit fabric. This combination of properties provides it with the highest level of cleanliness, abrasion resistance, and chemical compatibility.
The lightweight, tight-knit construction ensures low abrasion, which limits picking and snagging. It is critically low in particles, fibers, ions, and extractables.
Certifications: It is an ultrasonically sealed edge cleanroom laundered wiper required for ISO Class 3 and above environments.