Accessories for Force EXP Guard Column Cartridges, Restek
10118-320
: Restek
- Description:EXP Direct Connect Holder for EXP Guard Cartridges, Includes Fitting and Ferrules, Max. pressure: 20,000 psi (1,400 bar)
- Environmentally Preferable:
- Cat. No.:10118-320
- Supplier no.:25808