ContactUsLinkComponent
Thermal Conductivity Detector (TCD) 1/8" PPC Makeup Kit for PerkinElmer AutoSystem Series GCs, PerkinElmer
Thermal Conductivity Detector (TCD) 1/8" PPC Makeup Kit for PerkinElmer AutoSystem Series GCs, PerkinElmer
Catalog # 10138-730
Supplier:  PerkinElmer
Thermal Conductivity Detector (TCD) 1/8" PPC Makeup Kit for PerkinElmer AutoSystem Series GCs, PerkinElmer
Catalog # 10138-730
Supplier:  PerkinElmer
Supplier Number:  N6120150
Specifications
  • Description:
    Thermal Conductivity Detector (TCD) 1/8" PPC Makeup Kit
  • Cat. No.:
    10138-730
  • Supplier no.:
    N6120150
Specifications
About this item
Includes parts required to add PPC controlled make-up gas to an existing Thermal Conductivity Detector (TCD).
Ordering information: Requires PPC capability in PerkinElmer AutoSystem XL GC. Includes tubing, tee-piece, PPC pneumatics module and PPC frit #4 (N6120155).