ContactUsLinkComponent
Replacement O-Ring for VCO O-Ring Face Seal Fittings, Swagelok®, Restek
Replacement O-Ring for VCO O-Ring Face Seal Fittings, Swagelok®, Restek
  10855-412
 :  Restek
Replacement O-Ring for VCO O-Ring Face Seal Fittings, Swagelok®, Restek
  10855-412
 :  Restek
 :  23208
  • Description:
    Replacement O-Ring, 70 Durometer Fluorocarbon FKM, VCO 1/4", Size 010
  • Material:
    Stainless steel
  • Cat. No.:
    10855-412
  • Supplier no.:
    23208