Swagelok® VCO O-Ring Face Seal Fittings, Treated, Restek
Supplier: Restek
Certificates
About this item
Swagelok VCO O-ring face seal fittings are designed for rapid assembly in pipe, tube, and welded systems.
- Unique design allows easy installation where space is limited
- Assemblies can be used across a wide range of temperatures
- Smooth finish on gland face ensures positive seal
- Sealing is accomplished with a captive O-ring in the body component
Assemblies can be used from high pressure to critical vacuum across a wide range of temperatures.
Specifications
- Catalog No:
- 10854-754
- 10854-756
- 10855-524
- 10855-584
- Max. Operating Pressure:
- Up to 15,400 psig (1,061 bar)
- Up to 15,400 psig (1,061 bar)
- Up to 15,400 psig (1,061 bar)
- Up to 15,400 psig (1,061 bar)
- Max. Temperature:
- Up to 400 °F (204 °C)
- Up to 400 °F (204 °C)
- Up to 400 °F (204 °C)
- Up to 400 °F (204 °C)