Anti-beta Amyloid Mouse Monoclonal Antibody [clone: 6F/3D]
Catalog # 101982-364
Supplier: Avantor
Specifications
- Antibody Type:Primary
- Antigen Name:beta Amyloid
- Clonality:Monoclonal
- Clone:6F/3D
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:1 ml
- Format:Lyophilized tissue culture supernatant containing 15mM NaN3. Reconstitute with 1 ml of sterile distilled water.
- Storage Temperature:Store at 4 deg C until reconstitution. Following reconstitution aliquot and freeze at -20 deg C for long term storage. Avoid repeated freeze-thaw cycles.
- Immunogen:Beta amyloid antibody was raised in mouse using a sythetic peptide representing a site on the beta amyloid molecule as the immunogen.
- Cat. No.:101982-364
- Supplier no.:10RB113A
Specifications
About this item
Beta amyloid is an extracellular filamentous protein deposit found in the brain. It is the major protein component of amyloid cores, neuritic plaques and is also found as a deposit in neurofibrillary tangles. Alzheimer’s diseased brain tissue is characterised by an abundance of neurofibrillary tangles, neuropil threads and abnormal neuritis in senile plaques. Neurofibrillary tangles represent dense accumulations of ultrastructurally distinct paired helical filaments whose major component is a microtubule-associated tau protein.
IHC: 1:50
Type: Primary
Antigen: beta Amyloid
Clonality: Monoclonal
Clone: 6F/3D
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human