Anti-PTDSR Rabbit Polyclonal Antibody
102030-728
: Avantor
- Antibody Type:Primary
- Antigen Symbol:PTDSR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western Blot:Yes
- Size:50 µg
- Format:Lyophilized powder. Add 50 ul of distilled water. Final antibody concentration is 1 mg/ml in PBS buffer.
- Storage Temperature:Store at 4 deg C, following reconstitution, aliquot and store at -20 deg C. Avoid repeated freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:PTDSR antibody was raised in rabbit using the middle region of PTDSR as the immunogen
- Cat. No.:102030-728
- Supplier no.:70R7880
PTDSR is required during embryogenesis and differentiation of multiple organs during embryogenesis. PTDSR probably acts as a key regulator of hematopoietic differentiation. PTDSR may not be required for apoptotic cell clearance by macrophages but seems to be necessary for the regulation of macrophage cytokine responses
WB: 0.2-1 ug/ml
Type: Primary
Antigen: PTDSR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: