Anti-CD28 Mouse Monoclonal Antibody [clone: 15E8]
Catalog # 101982-778
Supplier: Avantor
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD28
- Clonality:Monoclonal
- Clone:15E8
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:100 µl
- Format:Liquid in 20mM TRIS, with 150mM NaCl, and 0.001% merthiolate.
- Storage Temperature:Aliquot and store at -20 deg C. Avoid repeated freeze/thaw cycles
- Immunogen:CD28 antibody was raised in mouse using human T lymphocytes as the immunogen.
- Cat. No.:101982-778
- Supplier no.:10RCD28BHU
Specifications
About this item
CD28 (Cluster of Differentiation 28) is one of the molecules expressed on T cells that provide co-stimulatory signals, which are required for T cell activation. CD28 is the receptor for B7.1 (CD80) and B7.2 (CD86). When activated by Toll-like receptor ligands, the B7.1 expression is upregulated in antigen presenting cells (APCs). The B7.2 expression on antigen presenting cells is constitutive. CD28 is the only B7 receptor constitutively expressed on naive T cells.
Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: 15E8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human