Anti-BRS3 Rabbit Polyclonal Antibody
Catalog # 101986-546
Supplier: Avantor
Specifications
- Antibody Type:Primary
- Antigen Name:Bombesin Receptor 3
- Antigen Symbol:BRS3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Size:50 µg
- Format:Purified by Immunoaffinity Chromatography and supplied in PBS with 0.1% NaN3.
- Antigen Synonyms:BRS3.|Bombesin receptor subtype 3|Bombesin like receptor 3|BRS 3|BB3
- Storage Temperature:Store at 4 deg C for short term storage. Aliquot and store at -70 deg C for long term storage. Avoid repeated freeze/thaw cycles.
- Concentration:1.0 mg/ml
- Immunogen:BRS3 antibody was raised in rabbit using a synthetic peptide conjugated to KLH as the immunogen.
- Cat. No.:101986-546
- Supplier no.:20RBR011
Specifications
About this item
The bombesin receptor subtype 3 also known as BRS-3 or BB3 is a protein which in humans is encoded by the BRS3 gene. Mammalian bombesin-like peptides are widely distributed in the central nervous system as well as in the gastrointestinal tract, where they modulate smooth-muscle contraction, exocrine and endocrine processes, metabolism, and behavior. They bind to G protein-coupled receptors on the cell surface to elicit their effects. BB3 is a G protein-coupled receptor.
IHC-P: 10 ug/ml
Type: Primary
Antigen: BRS3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human