To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Symbol:AAV VP1 + VP2 + VP3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:
- Western Blot:Yes
- Size:100 µl
- Environmentally Preferable:
- Format:Affinity purified IgG supplied in PBS with 0.02% NaN3.
- Storage Temperature:Store at 4 deg C for short term storage. Aliquot and store at -20 deg C for long term storage. Avoid repeated freeze/thaw cycles.
- Concentration:1.0 mg/ml
- Shipping Temperature:*Blue Ice*
- Immunogen:AAV VP1, VP2 & VP3 antibody was raised in rabbit using residues 75-86 [TPWGYFDFNRFHC] of major coat protein VP3 [AAV 2], residues 278-289 of capsid protein VP1 [AAV 6], and residues 277-288 of capsid AAV 3] as the immunogen.
- Cat. No.:102034-982
- Supplier no.:70RAR001
Adeno-associated virus (AAV) is a small virus which infects humans and some other primate species. AAV is not currently known to cause disease and consequently the virus causes a very mild immune response. AAV can infect both dividing and non-dividing cells and may incorporate its genome into that of the host cell.
WB: 1:500-1:1,000
Type: Primary
Antigen: AAV VP1 + VP2 + VP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: