To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Symbol:AAV (intact)
- Clonality:Monoclonal
- Clone:A20
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG3
- Reactivity:
- Size:50 µg
- Environmentally Preferable:
- Neutralisation:Yes
- Format:Lyophilizied from a solution containing 0.05% BSA, and 0.09% NaN3.
- Storage Temperature:Store at 4 deg C
- Shipping Temperature:*Blue Ice*
- Immunogen:AAV antibody (intact) was raised in mouse using adeno-associated virus caspid proteins and virus particles as the immunogen.
- Cat. No.:101982-260
- Supplier no.:10RA110A
Adeno-associated virus (AAV) is a small virus which infects humans and some other primate species. AAV is not currently known to cause disease and consequently the virus causes a very mild immune response. AAV can infect both dividing and non-dividing cells and may incorporate its genome into that of the host cell. These features make AAV a very attractive candidate for creating viral vectors for gene therapy, and for the creation of isogenic human disease models.
IF: 1:20, IP: 1:5
Type: Primary
Antigen: AAV (intact)
Clonality: Monoclonal
Clone: A20
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: