ContactUsLinkComponent
Rotational Vise
Catalog # 470228-934
undefined
Rotational Vise
Catalog # 470228-934
Supplier Number:  83-1117-00
Specifications
  • Description:
    Rotational vise
  • Cat. No.:
    470228-934
Specifications
About this item
Holds your projects in place at any angle.
  • Portable and light-weighted
Aluminum clamp-on vise. Swivel head with boll pivot allows ±45° vertical tilt and 360° swivel.