Mixed Calibration Standard, PerkinElmer
Catalog # 97011-960
Supplier: PerkinElmer
CAS Number:
Specifications
- Matrix:2% HNO₃
- Description:Mixed Calibration Standard
- Size:500 ml
- Cat. No.:97011-960
- Contents:50 µg/mL: As, K
10 µg/mL: La, Li, Mn, Ni, Sr, Zn
1 µg/mL: Ba, Mg - Supplier no.:N0691579