Trays, DWK Life Sciences
Tray for 100 (12 x 32 mm), 2 mL vials
Supplier: DWK Life Sciences (KIMBLE)
Supplier #: 2ML-TRAYSET
Jump to:
Product Details & Documents
Trays with lid.
Documents
Specifications
- Description:Tray for 100 (12 x 32 mm), 2 mL vials
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



