Weller® Contact Pyrometer, Apex
37697-504
- Description:Contact Pyrometer
- Cat. No.:37697-504
- Supplier no.:CP01
Contact pyrometer with large 20-gauge wire for use with WA2000 soldering iron analyzer.
- Quickly measures the temperature stability of a soldering iron tip