Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle
- Description:Diphtheria Toxin
- Size:1 mg
- Cat. No.:80055-232
- Supplier no.:322326-1MG
Specifications
About this item
Diphtheria toxin Catalyzes ADP-ribosylation of eukaryotic aminoacyltransferase II (EF2) using NAD as substrate, thereby inhibiting protein synthesis
- Induce internucleosomal breakdown
- It can cause DNA fragmentation and cytolysis in U937 cells
- Activation requires nicking with a protease followed by reduction with DTT
- Storage: +2 °C...+8 °C
For assays employing very low concentrations of diphtheria toxin, the use of a carrier protein, such as BSA or HSA, is recommended.The primary target is Eukaryotic aminoacyltransferase II (EF2)
This toxin is from Corynebacterium diphtheriae. This toxin is highly harmful