Method 535 Internal Standard (Butachlor ESA sodium salt), Restek
Catalog # 75835-168
Supplier: Restek
CAS Number:
Specifications
- Concentration:100 µg/ml in methanol
- Description:Butachlor ESA sodium salt
- Size:1 ml/Ampule
- Cat. No.:75835-168
- Supplier no.:33202