Order Entry
Puerto Rico
ContactUsLinkComponent
Hematoxylin 5% in SD alcohol
Flammable
Health
Hematoxylin 5% in SD alcohol
Catalog # 101410-916
Minority or Woman-Owned Business Enterprise
CAS Number:  
undefined
Flammable
Health
Hematoxylin 5% in SD alcohol
Catalog # 101410-916
Supplier Number:  26042-5
Minority or Woman-Owned Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Bottle
  • Size:
    500 ml
  • Cat. No.:
    101410-916
  • Supplier no.:
    26042-5

Specifications

About this item

 :
 :302.28 g/mol
Boiling Pt:78 °C (172 °F)
Flash Pt:13 °C (55 °F)
UN:1993
ADR:3,II
 :GHS02
 :GHS08
CAS Number:517-28-2
 :208-237-3
Formula:C₁₆H₁₄O₆
MDL Number:MFCD00078111