Order Entry
Puerto Rico
ContactUsLinkComponent
FAME #13 Mix (14 components), Restek
FAME #13 Mix (14 components), Restek
Catalog # RK35034
Supplier:  Restek
CAS Number:  
undefined
FAME #13 Mix (14 components), Restek
Catalog # RK35034
Supplier:  Restek
Supplier Number:  35034
CAS Number:  

Specifications

  • Concentration:
    Neat
  • Description:
    Fatty acid methyl ester, FAME Std #13
  • Size:
    50 mg
  • Cat. No.:
    RK35034
  • Supplier no.:
    35034

Specifications

About this item

Contains: Chain, Compound (CAS#), % by Weight; C16:00 Methyl palmitate (112-39-0), 3%; C16:01 Methyl palmitoleate (1120-25-8), 1%; C18:00 Methyl stearate (112-61-8), 2%; C18:01 Methyl oleate (112-62-9), 20%; C18:02 Methyl linoleate (112-63-0), 15%; C18:03 Methyl linolenate (301-00-8), 10%; C20:00 Methyl arachidate (1120-28-1), 1%; C20:01 Methyl eicosenoate (20839-34-3), 10%; C20:02 Methyl eicosadienoate (2463-02-7), 2%; C22:00 Methyl behenate (929-77-1), 1%; C22:01 Methyl erucate (1120-34-9), 30%; C22:02 Methyl docosadienoate (61012-47-3), 2%; C24:00:00 Methyl lignocerate (2442-49-1), 1%; C24:01:00 Methyl nervonate (2733-88-2), 2%