FAME #21 Mix (7 components), Restek
Catalog # RK35027
Supplier: Restek
CAS Number:
Specifications
- Concentration:Neat
- Description:Fatty acid methyl ester, FAME #21
- Size:50 mg/Ampule
- Cat. No.:RK35027
- Supplier no.:35027
Specifications
About this item
This mixture can be used for quantification (AOCS Method CE 1a–13) and approximate the compositions of the types of oils like olive, teaseed, and neatsfoot oil.
This mix contains seven components such as C14:00 methyl myristate (2%), C16:00 methyl palmitate (30%), C16:01 methyl palmitoleate (3%), C18:00 methyl stearate (14%), C18:01 methyl oleate (41%), C18:02 methyl linoleate (7%), C18:03 methyl linolenate (3%).
% represents weight/weight% basis (minimum 50 mg/ampoule).