Order Entry
Puerto Rico
ContactUsLinkComponent
FAME #9 Mix (5 components), Restek
FAME #9 Mix (5 components), Restek
Catalog # RK35018
Supplier:  Restek
CAS Number:  
undefined
FAME #9 Mix (5 components), Restek
Catalog # RK35018
Supplier:  Restek
Supplier Number:  35018
CAS Number:  

Specifications

  • Concentration:
    Neat
  • Description:
    Fatty acid methyl ester, FAME Std #9
  • Size:
    50 mg
  • Cat. No.:
    RK35018
  • Supplier no.:
    35018

Specifications

About this item

Contains: Chain, Compound (CAS#), % by Weight; C16:00 Methyl palmitate (112-39-0), 20%; C17:00 Methyl heptadecanoate (1731-92-6), 20%; C18:00 Methyl stearate (112-61-8), 20%; C19:00 Methyl nonadecanoate (1731-94-8), 20%; C20:00 Methyl arachidate (1120-28-1), 20%