Order Entry
Puerto Rico
ContactUsLinkComponent
FAME #5 Mix (5 components), Restek
FAME #5 Mix (5 components), Restek
Catalog # RK35014
Supplier:  Restek
CAS Number:  
undefined
FAME #5 Mix (5 components), Restek
Catalog # RK35014
Supplier:  Restek
Supplier Number:  35014
CAS Number:  

Specifications

  • Concentration:
    Neat
  • Description:
    Fatty acid methyl ester, FAME Std #5
  • Size:
    50 mg
  • Cat. No.:
    RK35014
  • Supplier no.:
    35014

Specifications

About this item

Contains: Chain, Compound (CAS#), % by Weight; C16:01 Methyl palmitoleate (1120-25-8), 20%; C18:01 Methyl oleate (112-62-9), 20%; C20:01 Methyl eicosenoate (20839-34-3), 20%; C22:01 Methyl erucate (1120-34-9), 20%; C24:01:00 Methyl nervonate (2733-88-2), 20%