Order Entry
Puerto Rico
ContactUsLinkComponent
FAME #4 Mix (5 components), Restek
FAME #4 Mix (5 components), Restek
Catalog # RK35013
Supplier:  Restek
CAS Number:  
undefined
FAME #4 Mix (5 components), Restek
Catalog # RK35013
Supplier:  Restek
Supplier Number:  35013
CAS Number:  

Specifications

  • Concentration:
    Neat
  • Description:
    Fatty acid methyl ester, FAME Std #4
  • Size:
    50 mg
  • Cat. No.:
    RK35013
  • Supplier no.:
    35013

Specifications

About this item

Contains: Chain, Compound (CAS#), % by Weight; C16:00 Methyl palmitate (112-39-0), 20%; C18:00 Methyl stearate (112-61-8), 20%; C20:00 Methyl arachidate (1120-28-1), 20%; C22:00 Methyl behenate (929-77-1), 20%; C24:00:00 Methyl lignocerate (2442-49-1), 20%