Order Entry
Puerto Rico
ContactUsLinkComponent
531.2 Carbamate Pesticide Calibration Mixture (11 components), Restek
531.2 Carbamate Pesticide Calibration Mixture (11 components), Restek
Catalog # RK32435
Supplier:  Restek
CAS Number:  
undefined
531.2 Carbamate Pesticide Calibration Mixture (11 components), Restek
Catalog # RK32435
Supplier:  Restek
Supplier Number:  32435
CAS Number:  

Specifications

  • Concentration:
    100 µg/ml in acetonitrile
  • Description:
    531.2 Carbamate Pesticide Calibration Mixture
  • Size:
    1 ml/Ampule
  • Cat. No.:
    RK32435
  • Certification:
    CRM
  • Supplier no.:
    32435

Specifications

About this item

This mix contains 11 components.

  • Complete set of materials for N-methylcarbamoyloximes and N-methylcarbamates.
  • New mix satisfies latest update of EPA Method 531.2.
  • Formulated in acetonitrile for stability and convenience for LC analysis.

Because carbamates, especially aldicarb sulfone, are unstable, carbamates analyses usually are HPLC based. The U.S. EPA method for monitoring these compounds in drinking water, Method 531, calls for HPLC with post-column derivatization. 1-Naphthol, a fluorescent metabolite of carbaryl, was added to the analyte list in 2003. Our 531.2 Carbamate Pesticides Calibration Mixture includes 1-naphthol. For stability, we formulate the mix in acetonitrile.

Contains: Aldicarb (116-06-3); Aldicarb sulfone (1646-88-4); Aldicarb sulfoxide (1646-87-3); Carbaryl (Sevin) (63-25-2); Carbofuran (1563-66-2); 3-Hydroxycarbofuran (16655-82-6); Methiocarb (2032-65-7); Methomyl (16752-77-5); 1-Naphthol (90-15-3); Oxamyl (23135-22-0); Propoxur (Baygon) (114-26-1).

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.