Dieldrin, Restek
Catalog # RK32218
Supplier: Restek
CAS Number:
Specifications
- Concentration:1000 µg/ml in methanol
- CAS No.:60-57-1
- Description:Dieldrin
- Size:1 ml/Ampule
- Formula:C₁₂H₈Cl₆O
- Description:Dieldrin
- Cat. No.:RK32218
- Certification:CRM
- Supplier no.:32218
Specifications
About this item
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.