Order Entry
Puerto Rico
ContactUsLinkComponent
Pesticide Performance Evaluation Mix with Surrogates (8 components), Restek
Pesticide Performance Evaluation Mix with Surrogates (8 components), Restek
Catalog # RK32074
Supplier:  Restek
CAS Number:  
undefined
Pesticide Performance Evaluation Mix with Surrogates (8 components), Restek
Catalog # RK32074
Supplier:  Restek
Supplier Number:  32074
CAS Number:  

Specifications

  • Concentration:
    In hexane
  • Description:
    Pesticide Performance Evaluation Mix w/Surrogates
  • Size:
    1 ml/Ampule
  • Cat. No.:
    RK32074
  • Certification:
    CRM
  • Supplier no.:
    32074

Specifications

About this item

This mix contains eight components.

Components are α-BHC (1 μg/ml), β-BHC (1 μg/ml), γ-BHC (1 μg/ml), 4,4'-DDT (10 μg/ml), decachlorobiphenyl (SS) (2 μg/ml), endrin (5 μg/ml), methoxychlor (25 μg/ml), 2,4,5,6-tetrachloro-m-xylene (2 μg/ml).

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.