Herbicide Mix #4 (8 components), Derivatized Form, Restek
Catalog # RK32062
Supplier: Restek
CAS Number:
Specifications
- Concentration:200 µg/ml each in hexane
- Description:Herbicide Mix #4
- Size:1 ml/Ampule
- Cat. No.:RK32062
- Certification:CRM
- Supplier no.:32062
Specifications
About this item
This mix contains eight components.
Components are acifluorfen methyl ester, bentazon methyl ester, chloramben methyl ester, DCPA methyl ester (chlorthal-dimethyl), 3,5-dichlorobenzoic acid methyl ester, 4-nitroanisole, pentachloroanisole, picloram methyl ester.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.