Order Entry
Puerto Rico
ContactUsLinkComponent
508 Performance Check Mix (4 components), Restek
508 Performance Check Mix (4 components), Restek
Catalog # RK32045
Supplier:  Restek
CAS Number:  
undefined
508 Performance Check Mix (4 components), Restek
Catalog # RK32045
Supplier:  Restek
Supplier Number:  32045
CAS Number:  

Specifications

  • Concentration:
    In methyl tert-butyl ether
  • Description:
    508 Performance Check Mix
  • Size:
    1 ml/Ampule
  • Cat. No.:
    RK32045
  • Certification:
    CRM
  • Supplier no.:
    32045

Specifications

About this item

This mix contains four components.

Components are δ-BHC (0.4 μg/ml), chlorothalonil (0.5 μg/ml), chlorpyrifos (0.02 μg/ml), DCPA methyl ester (Chlorthal-dimethyl) (0.5 μg/ml).

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.