Order Entry
United States
ContactUsLinkComponent
8270 Calibration Kit, Restek
8270 Calibration Kit, Restek
Catalog # RK31626
Supplier:  Restek
CAS Number:  
undefined
8270 Calibration Kit, Restek
Catalog # RK31626
Supplier:  Restek
Supplier Number:  31626
CAS Number:  

Specifications

  • Concentration:
    Contains 1 ml each of these mixtures
  • Description:
    8270 Calibration Kit
  • Size:
    Kit
  • Environmentally Preferable:
  • Cat. No.:
    RK31626
  • Certification:
    CRM
  • Supplier no.:
    31626

Specifications

About this item

Contains 1 ml each of the followig mixtures, 8270 calibration mix #1, 8270 calibration mix #2, 8270 calibration mix #3, 8270 calibration mix #4, 8270 calibration mix #5 revised and 3-methylcholanthrene standard.

Contains: 31618: 8270 Calibration Mix #1; Benzoic acid (65-85-0); 4-Chloro-3-methylphenol (59-50-7); 2-Chlorophenol (95-57-8); 2,4-Dichlorophenol (120-83-2); 2,6-Dichlorophenol (87-65-0); 2,4-Dimethylphenol (105-67-9); 4,6-Dinitro-2-methylphenol (Dinitro-o-cresol) (534-52-1); 2,4-Dinitrophenol (51-28-5); Dinoseb (88-85-7); 2-Methylphenol (o-cresol) (95-48-7); 3-Methylphenol (m-cresol) (108-39-4); 4-Methylphenol (p-cresol) (106-44-5); 2-Nitrophenol (88-75-5); 4-Nitrophenol (100-02-7); Pentachlorophenol (87-86-5); Phenol (108-95-2); 2,3,4,6-Tetrachlorophenol (58-90-2); 2,4,5-Trichlorophenol (95-95-4); 2,4,6-Trichlorophenol (88-06-2);31619: 8270 Calibration Mix #2; Aniline (62-53-3); Benzidine (92-87-5); 4-Chloroaniline (106-47-8); 3,3'-Dichlorobenzidine (91-94-1); Diphenylamine (122-39-4)*; 2-Nitroaniline (88-74-4); 3-Nitroaniline (99-09-2); 4-Nitroaniline (100-01-6); N-Nitrosodimethylamine (62-75-9); N-Nitrosodi-n-propylamine (621-64-7); Pyridine (110-86-1);31620: 8270 Calibration Mix #3; Aramite (140-57-8); Bis(2-chloroethyl)ether (111-44-4); Bis(2-chloroethoxy)methane (111-91-1); 4-Bromophenyl phenyl ether (101-55-3); Chlorobenzilate (510-15-6); 2-Chloronaphthalene (91-58-7); 4-Chlorophenyl phenyl ether (7005-72-3); 1,2-Dichlorobenzene (95-50-1); 1,3-Dichlorobenzene (541-73-1); 1,4-Dichlorobenzene (106-46-7); 1,3-Dinitrobenzene (99-65-0); Hexachlorobenzene (118-74-1); Hexachlorobutadiene (87-68-3); Hexachlorocyclopentadiene (77-47-4); Hexachloroethane (67-72-1); Hexachloropropene (1888-71-7); Isodrin (465-73-6); Kepone (143-50-0); 2,2'-Oxybis(1-chloropropane) (108-60-1); Pentachlorobenzene (608-93-5); Pentachloronitrobenzene (quintozene) (82-68-8); 1,2,4,5-Tetrachlorobenzene (95-94-3); 1,2,4-Trichlorobenzene (120-82-1);31621: 8270 Calibration Mix #4; Acetophenone (98-86-2); Azobenzene (103-33-3)*; Benzyl alcohol (100-51-6); Bis(2-ethylhexyl)phthalate (117-81-7); Benzyl butyl phthalate (85-68-7); Dibenzofuran (132-64-9); Diethylphthalate (84-66-2); Dimethylphthalate (131-11-3); Di-n-butyl phthalate (84-74-2); Di-n-octyl phthalate (117-84-0); 2,4-Dinitrotoluene (121-14-2); 2,6-Dinitrotoluene (606-20-2); Ethyl methanesulfonate (62-50-0); Isophorone (78-59-1); Isosafrole (cis & trans) (120-58-1); Methyl methanesulfonate (66-27-3); 1,4-Naphthoquinone (130-15-4); Nitrobenzene (98-95-3); 4-Nitroquinoline-N-oxide (56-57-5); Phenacetin (62-44-2); Safrole (94-59-7); 1,3,5-Trinitrobenzene (99-35-4);31995: 8270 Calibration Mix #5, Revised; Acenaphthene (83-32-9); Acenaphthylene (208-96-8); Anthracene (120-12-7); Benz(a)anthracene (56-55-3); Benzo(a)pyrene (50-32-8); Benzo(b)fluoranthene (205-99-2); Benzo(ghi)perylene (191-24-2); Benzo(k)fluoranthene (207-08-9); Chrysene (218-01-9); Dibenz(a,h)anthracene (53-70-3); Fluoranthene (206-44-0); Fluorene (86-73-7); Indeno(1,2,3-cd)pyrene (193-39-5); 1-Methylnaphthalene (90-12-0); 2-Methylnaphthalene (91-57-6); Naphthalene (91-20-3); Phenanthrene (85-01-8); Pyrene (129-00-0);31996: 3-Methylcholanthrene Standard; 3-Methylcholanthrene (56-49-5).

Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.